Statfree® High Density Conductive Foam, Desco
Catalogue # 89131-086
Fournisseur: Desco
Product available on GSA Advantage®
Spécifications
- Description:Statfree® high density conductive foam
- Dimensions:0.75×24×36"
- Cat. no.:89131-086
- Supplier No.:12550
Spécifications
À propos de cet article
High Density Conductive Foam is antistatic and low tribocharging, which minimizes charge generation.
- Antistatic, low tribo charging
- Dissipative and corrosion resistant
- Uniform protection
- Volume Resistance: 10E3 – 10E5 ohms
- Surface Resistance: 10E3 – 10E5 ohms
- Made in the United States of America
Antistatic, low tribocharging minimizes charge generation; material will conduct charges if grounded. Corrosion resistant design is ideal for long-term storage. Uniform protection will not affect the solderability of device leads.
Certifications: Lead-free, RoHS compliant.