Snap Kits, Desco | Avantor
Snap Kits, Desco
Supplier:   Desco
Snap Kits, Desco
Supplier:  Desco
Certificates
Snap Kits, Desco
Snap Kits, Desco
2 Options Available Below
Product Details & Documents
Snaps are user installable and allow use precisely where needed.
  • Snap is user installable
  • ESD common Point ground symbol label
  • Made in The United States of America
ESD Common Point Ground Symbol Label used to indicate the location of an acceptable common point ground as defined by ANSI/EOS-ESD-S6.1. The Push and Clinch Snap Socket kit includes pronged socket snap and common point ground symbol label. The Universal Snap kit includes 10 mm stud, 10 mm socket, long screw, and short screw.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Push and Clinch Snap Socket
    10 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4.08
    $4.08
    Each
       $4.08
    Supplier Number: 09863
  • Universal Snap Kit
    10 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6.42
    $6.42
    Each
       $6.42
    Supplier Number: 09864