Hei-VAP Ultimate Rotary Evaporators, Heidolph®
Supplier: Heidolph NA, LLC
Certificates
About this item
Designed as precise setting of rotation speed and heating bath temperature on the digital touch display or for quick access during an active process, directly via the knobs on the operating panel.
- Clear display of all values
- Central operating panel with 7" touch display
- Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
- Setting of values either via the touch display or the separate quick access knobs
- For precise monitoring, the pictogram on the display also symbolizes active functions
- LED ring lights and pictogram to display the activity even from a distance
- USB and Micro SD plus LAN / RS 232 interfaces
- Optional display of vapor temperature
- Define and save own evaporation processes (ramps)
It has Separate knobs for rotation and heating with locking function to prevent accidental readjusting, with integrated solvent library to facilitate adjustment of the required vacuum on an external vacuum controller. Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation.
Ordering information: G1 glassware set is a diagonal coil condenser. G3 glassware set is a vertical coil condenser. G3 XL glassware is a higher evaporation rate condensor. Glassware set G5 is a dry ice condenser. Glassware set G6 is a vertical condenser.
Specifications
- Catalog No:
- 76338-400
- 76338-416
- 76338-438
- 76338-436
- 76338-414
- 76338-420
- 76338-418
- 76338-408
- 76338-430
- 76338-406
- 76338-428
- 76338-412
- 76338-434
- 76338-410
- 76338-432
- 76338-422
- 76338-404
- 76338-426
- 76338-402
- 76338-424
- Model:
- Hei-VAP Ultimate, HL, G1
- Hei-VAP Ultimate, HL, G5 B
- Hei-VAP Ultimate, ML, G6 B
- Hei-VAP Ultimate, ML, G5 B
- Hei-VAP Ultimate, HL, G 3B XL
- Hei-VAP Ultimate, ML, G1
- Hei-VAP Ultimate, HL, G6 B
- Hei-VAP Ultimate, HL, G6
- Hei-VAP Ultimate, ML, G1B
- Hei-VAP Ultimate, HL, G5
- Hei-VAP Ultimate, ML, G6
- Hei-VAP Ultimate, HL, G 3B
- Hei-VAP Ultimate, ML, G3 B XL
- Hei-VAP Ultimate, HL, G1B
- Hei-VAP Ultimate, ML, G3 B
- Hei-VAP Ultimate, ML, G3
- Hei-VAP Ultimate, HL, G3 XL
- Hei-VAP Ultimate, ML, G5
- Hei-VAP Ultimate, HL, G3
- Hei-VAP Ultimate, ML, G3 XL
- Rotation speed:
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- Heating capacity:
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- Cooling surface:
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 2200 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 2200 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 2200 cm²
- 1400 cm²
- 1400 cm²
- 2200 cm²
- Bath capacity:
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- Protection class DIN EN 60529:
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- Timer:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature range:
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- Max. flask capacity:
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- Bath Ø:
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- Accuracy:
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C