Hei-VAP Expert Rotary Evaporators, Heidolph®
Fournisseur: Heidolph NA, LLC
Certificats
À propos de cet article
Designed as precise setting of rotation speed and heating bath temperature on the digital touch display or for quick access during an active process, directly via the knobs on the operating panel.
- Clear display of all values
- Central operating panel with 5" touch display
- Separate knobs for rotation and heating with locking function to prevent accidental readjusting
- LED ring lights and pictogram to display the activity even from a distance
- USB and Micro SD interfaces
- Optional display of vapor temperature
- Subsequent upgrade to Hei-VAP Expert Control possible, including vacuum and chiller control
It has integrated solvent library to facilitate adjustment of the required vacuum on an external vacuum controller with timer, limitation of maximum temperature, multilingual menu and text display for error messages. Available in Hand Lift (HL) and Motor Lift (ML) Configurations.
Ordering information: G1 glassware set is a diagonal coil condenser. G3 glassware set is a vertical coil condenser. G3 XL glassware is a higher evaporation rate condensor. Glassware set G5 is a dry ice condenser. Glassware set G6 is a vertical condenser.
Spécifications
- Catalog No:
- 76338-320
- 76338-336
- 76338-358
- 76338-356
- 76338-334
- 76338-340
- 76338-338
- 76338-350
- 76338-328
- 76338-348
- 76338-326
- 76338-354
- 76338-332
- 76338-330
- 76338-352
- 76338-342
- 76338-346
- 76338-324
- 76338-344
- 76338-322
- Model:
- Hei-VAP Expert, HL, G1
- Hei-VAP Expert, HL, G5 B
- Hei-VAP Expert, ML, G6 B
- Hei-VAP Expert, ML, G5 B
- Hei-VAP Expert, HL, G 3B XL
- Hei-VAP Expert, ML, G1
- Hei-VAP Expert, HL, G6 B
- Hei-VAP Expert, ML, G1B
- Hei-VAP Expert, HL, G6
- Hei-VAP Expert, ML, G6
- Hei-VAP Expert, HL, G5
- Hei-VAP Expert, ML, G3 B XL
- Hei-VAP Expert, HL, G 3B
- Hei-VAP Expert, HL, G1B
- Hei-VAP Expert, ML, G3 B
- Hei-VAP Expert, ML, G3
- Hei-VAP Expert, ML, G5
- Hei-VAP Expert, HL, G3 XL
- Hei-VAP Expert, ML, G3 XL
- Hei-VAP Expert, HL, G3
- Rotation speed:
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- Heating capacity:
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- 1300 W
- Cooling surface:
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 2200 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 2200 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 1400 cm²
- 2200 cm²
- 2200 cm²
- 1400 cm²
- Bath capacity:
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- Protection class DIN EN 60529:
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- IP 20
- Timer:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature range:
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- Max. flask capacity:
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- Bath Ø:
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- Accuracy:
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C