RACK’N’LAB Sample Bag Carriers, Labplas | Avantor
RACK’N’LAB Sample Bag Carriers, Labplas
Supplier:   Labplas
RACK’N’LAB Sample Bag Carriers, Labplas
Supplier:  Labplas
Certificates
RACK’N’LAB Sample Bag Carriers, Labplas
RACK’N’LAB Sample Bag Carriers, Labplas
2 Options Available Below
Product Details & Documents
The RACK’N’LAB is best used to transport bags used in food testing and environmental water sampling amongst many other industries. Its innovative design allows multiple bags to be used whilst being held upright without the need for bottles or specifically designed bags.
  • Made of FDA approved virgin polypropylene
  • Shipped in one inch thick boxes with three flat racks inside
  • Assembled or dismantled in less than one minute
  • The sturdy handle facilitates safe transportation
  • Identified rows allow for easy identification of samples
  • Up to 40 ml can be retained in case of a leak
  • Racks are stackable and will easily fit in standard coolers
This light weight and versatile rack takes up very little storage space when stored in its flat form and is available in two different sizes.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Sample bag carrier
    305×247×117 mm (12×9.72×4.6")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $959.59
    $959.59
    Case of 1
       $959.59
    Supplier Number: SA5-7012
  • Sample bag carrier
    305×171×76 mm (12×6.75×3")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $797.00
    $797.00
    Case of 1
       $797.00
    Supplier Number: SA5-3070