Capinsert™ Color Coding Inserts, Simport Scientific
Capinsert™ Color Coding Inserts, Simport Scientific
Supplier:   Simport Scientific
Capinsert™ Color Coding Inserts, Simport Scientific
Supplier:  Simport Scientific
Certificates
Capinsert™ Color Coding Inserts, Simport Scientific
Capinsert™ Color Coding Inserts, Simport Scientific
5 Options Available Below
Product Details & Documents
Capinserts™ can be used for all color coding needs.
  • Made of polypropylene
  • With write-on frosted area
  • Available in ten colors
The Capinsert™ is used to color code a multitude of Simport® products according to your specific needs. It is inserted on top of the closure and has a write-on frosted area for sample identification.
Product Family Options
Product Information
  • Désignation
    Color
    Availability
    Price
  • Cap Insert
    Blue
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $41.11
    $41.11
    Case of 500
       $41.11
    Supplier Number: T345B
  • Cap Insert
    Lilac
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $41.11
    $41.11
    Case of 500
       $41.11
    Supplier Number: T345L
  • Cap Insert
    Red
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $41.11
    $41.11
    Case of 500
       $41.11
    Supplier Number: T345R
  • Cap Insert
    White
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $41.11
    $41.11
    Case of 500
       $41.11
    Supplier Number: T345W
  • Cap Insert
    Yellow
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $40.79
    $40.79
    Case of 500
       $40.79
    Supplier Number: T345Y