Fiber Glass Reinforced Containers, Molded Fiber Glass Tray Company
Supplier: Molded Fiber Glass Tray
Certificates
About this item
These containers possess superior physical properties (temperature rating, weight capacity, chemical resistance) over competing thermoplastic products.
- Design saves space - nests when empty
- Can be used as a drawer
- Freezer safe
They are ideal for storage of small parts.
Specifications
- Catalog No:
- 89022-812
- 89022-822
- 89022-820
- 89022-818
- 89022-816
- pH range:
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- Temperature range:
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently