Kimtech™ A8 Zippered Lab Coats, Kimberly-Clark Professional
Supplier: Microflex
Certificates
About this item
Kimtech™ A8 Zippered Lab Coats are made with a protective three-layer SMS fabric and are recommended where there is the potential for light fluid contact.
- Recommended for use in areas with a potential for light fluid contact
- Recyclable via RightCycle by Kimberly-Clark Professional
This protective lab coat features a high mandarin collar, soft knit cuffs, zippered front closure to minimize contamination risk, and no pockets.
Packaging: Double bagged.
Specifications
- Catalog No:
- 76339-814
- 76339-816
- 76339-818
- 76339-820
- 76339-822
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory