TruCLEAN® Adjustable Polymer Mop Handle, Perfex
TruCLEAN® Adjustable Polymer Mop Handle, Adjustable from 108 - 216" (9 - 18')
Catalog # 89095-998
Supplier: Perfex
Supplier #: 88-18
Jump to:
Product Details & Documents
Optimize working posture, improve comfort level, and clean more efficiently.
- High-quality, durable polymer structure
- Super-smooth finish prevents microbial growth
- Non-conductive, chemically resistant
- Autoclave sterilization compatible
TruCLEAN® Adjustable Mop Handle is designed for compliance with the critical cleaning standards of today's controlled environments. Lightweight and durable handle, manufactured with high-quality polymer and composite materials using pull-wound technology.
Comfortable 1" diameter shaft reduces strain while transferring maximum energy with minimum exertion. Super-smooth finish prevents microbial growth. Compatible with gamma, ETO, and autoclave sterilization.
Specifications
- Désignation:TruCLEAN® Adjustable Polymer Mop Handle
- Longueur:Adjustable from 108 - 216" (9 - 18')