Colorant selon Wright | Avantor
Colorant selon Wright
Colorant selon Wright
Supplier:  Azer Scientific
Certificates
Colorant selon Wright
2 Options Available Below
Product Details & Documents
Wright stain is a polychromatic hematology stain consisting of eosin and methylene blue.
  • Dye mixture produces multiple colors to clearly differentiate white blood cells, red blood cells appear red-orange
  • Suitable for automated and manual staining protocols
  • Wright stain can be used for urine cytology and fine needle aspirate cell preparations
Synonymes:  Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
  • CAS Number:68988-92-1
  • Numéro MDL:MFCD00082143
Product Family Options
Informations sur le produit
  • Taille
    Availability
    Price
  • 1 gal.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2 (1/Itm/Bx)
    Case of 2 (1/Itm/Bx) - $1,132.76
    $1,132.76
    Case of 2 (1/Itm/Bx)
       $1,132.76
    Supplier Number: ES920
  • 32 oz.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (32oz)
    Each (32oz) - $1,299.60
    $1,299.60
    Each (32oz)
       $1,299.60
    Supplier Number: ES918