Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Dual Luer Swivel Fittings
- Cat. no.:89134-252
- Gauge:22G
- Supplier No.:QCS-D
The Quick Connect™ Swivel provides Luer-Lock fittings for trouble-free connection to extension lines and tethers, making connecting simple, fast, and easy.
- Fits all standard luer fittings
- Double Luer and Single Luer models available
- EtO exposed
Add injection caps, valves, or stopcocks to simplify dosing or sampling procedures. Part of SAI's Quick Connect™ System, which includes the Quick Connect™ Tether and Quick Connect™ Harness for tethered small animal infusion studies.