Distilling Receivers, Moisture Test, Overflow Tube, Dean-Stark, Chemglass
Distilling receivers feature a overflow tube.
- Overflow tube
- Between the uptake tube and the graduated receiver
Similar to CG-1258 but with the addition of an overflow tube between the uptake tube and the graduated receiver.
Frequently Bought Together