About this item
Tapered polyurethane rat arterial catheters are designed to enter the smaller, more muscular vessels while maintaining strength and durability at the externalization point.
- Tapered design improves fluid dynamics
- Suture beads secure the catheter for improved patency
- Small tip size and smooth surface improve the catheter implantation process
- Radio opaque for catheter placement verification
- Accessible with a 22G blunt needle
Both a tapered and a step-up option of this device are available.
For animal 150 g plus.
Tip O.D 2.3F, tip length 4 cm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.