ContactUsLinkComponent
Liquid-Liquid Extractor, Continuous, Round Bottom, Chemglass
Catalog # 80068-150
Supplier:  Chemglass
Small Business Enterprise
undefined
Liquid-Liquid Extractor, Continuous, Round Bottom, Chemglass
Catalog # 80068-150
Supplier:  Chemglass
Supplier Number:  CG-1366-01
Small Business Enterprise
Specifications
  • Description:
    Liquid-Liquid Extractor, Continous
  • Cat. no.:
    80068-150
  • Supplier No.:
    CG-1366-01
Specifications
About this item
Round bottom design has the return tube sealed into the bottom of the extractor for more efficient extractions.
  • For samples of 1 liter plus approximately 200 ml of extracting solvent
  • Extractor has a 45/50 top outer joint and a 24/40 inner joint on the uptake tube
Ordering information: Flask and condenser must be ordered separately.