Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Mouse Recombinant VEGF 165 (from E. coli)

50 µg
Catalog # 76303-664
Supplier:   Peprotech
 :  AF-450-32-50UG
Mouse Recombinant VEGF 165 (from <i>E. coli</i>)
Catalog # 76303-664
Supplier:  Peprotech
CAS Number:  
Mouse Recombinant VEGF 165 (from E. coli)

50 µg

CAS Number:  
Price
$1,464.75
Log in for availability
Each (50µG)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

VEGF is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothelial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays a prominent role in normal and pathological angiogenesis. Substantial evidence implicates VEGF in the induction of tumor metastasis and intra-ocular neovascular syndromes. VEGF signals through three receptors; fms-like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Recombinant Murine VEGF 165 is a 39.0 kDa, disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains.

  • Animal-free
Specifications

Specifications

  • Taille:
    50 µg
  • Protein/peptide name:
    VEGF 165
  • Protein synonyms:
    Vascular Endothelial Growth Factor165|VEGF-A|Glioma-derived endothelial cell mitogen|Folliculostellate cell-derived growth factor
  • Purity:
    98%
  • Endotoxin level:
    Low
  • Sequence:
    MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKHCEPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR
  • Source:
    E. coli
  • Biological activity:
    Determined by the dose-dependent stimulation of the proliferation of human umbilical vein endothelial cells (HUVEC) using a concentration range of 1.0-5.0 ng/ml.
  • Molecular weight:
    39 kDa
  • Shipping temperature:
    Ambient
  • Cat. No.:
    76303-664
  • Protein/peptide type:
    Recombinant
  • Gene ID:
    22339
  • Formulation:
    Lyophilized
  • Conjugation:
    Unconjugated
  • Species:
    Mouse
  • Animal-free:
    Y

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.