Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mouse Recombinant IL-10 (from E. coli) | Avantor

Mouse Recombinant IL-10 (from E. coli)

Supplier:   Peprotech
Mouse Recombinant IL-10 (from <i>E. coli</i>)
Supplier:  Peprotech
Certificates

6 Options Available Below

  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

IL-10 is an immunosuppressive cytokine produced by a variety of mammalian cell types including macrophages, monocytes, T cells, B cells and keratinocytes. IL-10 inhibits the expression of proinflammatory cytokines such as IL-1 and TNF-alpha. Like IL-4, IL-10 enhances humoral immune responses and attenuates cell-mediated immune reactions. Human IL-10 is active on murine cells, but murine IL-10 is inactive on human cells. Recombinant Murine IL-10 is an 18.7 kDa protein of 161 amino acid residues.

  • Animal-free
Specifications for Product Family

Spécifications

Spécifications

  • Catalog No:
  • 76303-582
  • 76304-128
  • 76304-130
  • 76304-132
  • 76304-134
  • 76303-600
  • Protein/peptide name:
  • IL-10
  • IL-10
  • IL-10
  • IL-10
  • IL-10
  • IL-10
  • Protein synonyms:
  • B-TCGF|TGIF|Interleukin-10|CSIF
  • B-TCGF|TGIF|Interleukin-10|CSIF
  • B-TCGF|TGIF|Interleukin-10|CSIF
  • B-TCGF|TGIF|Interleukin-10|CSIF
  • B-TCGF|TGIF|Interleukin-10|CSIF
  • B-TCGF|TGIF|Interleukin-10|CSIF
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • 16153
  • 16153
  • 16153
  • 16153
  • 16153
  • 16153
  • Sequence:
  • MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
  • MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
  • MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
  • MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
  • MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
  • MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
  • Animal-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Low
  • Biological activity:
  • Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
  • Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
  • Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
  • Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
  • Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
  • Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
  • Purity:
  • 98%
  • 98%
  • 98%
  • 98%
  • 98%
  • 98%
  • Formulation:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Molecular weight:
  • 18.7 kDa
  • 18.7 kDa
  • 18.7 kDa
  • 18.7 kDa
  • 18.7 kDa
  • 18.7 kDa
  • Shipping temperature:
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient

Spécifications

Product Family Options

Informations sur le produit

  • Taille
    Availability
    Price
  • 2 μg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (2µG)
    Each (2µG) - $262.86
    $262.86
    Each (2µG)
       $262.86
    La quantité minimale de commande est de 1

    Supplier Number: AF-210-10-2UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    Marketsource
    More
    Specifications
    Cat. No.
    76303-582
    Protein/peptide name
    IL-10
    Protein synonyms
    B-TCGF|TGIF|Interleukin-10|CSIF
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16153
    Sequence
    MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
    Animal-free
    Y
    Endotoxin level
    Low
    Biological activity
    Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
    Purity
    98%
    Formulation
    Lyophilized
    Molecular weight
    18.7 kDa
    Shipping temperature
    Ambient
    Marketsource
  • 10 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (10µG)
    Each (10µG) - $612.14
    $612.14
    Each (10µG)
       $612.14

    Supplier Number: AF-210-10-10UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    76304-128
    Protein/peptide name
    IL-10
    Protein synonyms
    B-TCGF|TGIF|Interleukin-10|CSIF
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16153
    Sequence
    MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
    Animal-free
    Y
    Endotoxin level
    Low
    Biological activity
    Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
    Purity
    98%
    Formulation
    Lyophilized
    Molecular weight
    18.7 kDa
    Shipping temperature
    Ambient
  • 100 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (100µG)
    Each (100µG) - $3,237.87
    $3,237.87
    Each (100µG)
       $3,237.87

    Supplier Number: AF-210-10-100UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    76304-130
    Protein/peptide name
    IL-10
    Protein synonyms
    B-TCGF|TGIF|Interleukin-10|CSIF
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16153
    Sequence
    MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
    Animal-free
    Y
    Endotoxin level
    Low
    Biological activity
    Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
    Purity
    98%
    Formulation
    Lyophilized
    Molecular weight
    18.7 kDa
    Shipping temperature
    Ambient
  • 250 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (250µG)
    Each (250µG) - $4,873.50
    $4,873.50
    Each (250µG)
       $4,873.50

    Supplier Number: AF-210-10-250UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    76304-132
    Protein/peptide name
    IL-10
    Protein synonyms
    B-TCGF|TGIF|Interleukin-10|CSIF
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16153
    Sequence
    MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
    Animal-free
    Y
    Endotoxin level
    Low
    Biological activity
    Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
    Purity
    98%
    Formulation
    Lyophilized
    Molecular weight
    18.7 kDa
    Shipping temperature
    Ambient
  • 500 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500µG)
    Each (500µG) - $8,328.02
    $8,328.02
    Each (500µG)
       $8,328.02

    Supplier Number: AF-210-10-500UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    76304-134
    Protein/peptide name
    IL-10
    Protein synonyms
    B-TCGF|TGIF|Interleukin-10|CSIF
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16153
    Sequence
    MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
    Animal-free
    Y
    Endotoxin level
    Low
    Biological activity
    Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
    Purity
    98%
    Formulation
    Lyophilized
    Molecular weight
    18.7 kDa
    Shipping temperature
    Ambient
  • 1 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1mg)
    Each (1mg) - $13,898.32
    $13,898.32
    Each (1mg)
       $13,898.32

    Supplier Number: AF-210-10-1MG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    76303-600
    Protein/peptide name
    IL-10
    Protein synonyms
    B-TCGF|TGIF|Interleukin-10|CSIF
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16153
    Sequence
    MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
    Animal-free
    Y
    Endotoxin level
    Low
    Biological activity
    Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
    Purity
    98%
    Formulation
    Lyophilized
    Molecular weight
    18.7 kDa
    Shipping temperature
    Ambient
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.