Human Recombinant IL-7 (from E. coli)
À propos de cet article
IL-7 is a hematopoietic growth factor that primarily affects early B and T cells. Produced by thymic stromal cells, spleen cells and keratinocytes, IL-7 can also co-stimulate the proliferation of mature T cells in combination with other factors, such as ConA and IL-2. Human and murine IL-7 are cross-species reactive. Recombinant Human IL-7 is a 17.4 kDa protein containing 153 amino acid residues.
- Animal-free
2 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
IL-7 is a hematopoietic growth factor that primarily affects early B and T cells. Produced by thymic stromal cells, spleen cells and keratinocytes, IL-7 can also co-stimulate the proliferation of mature T cells in combination with other factors, such as ConA and IL-2. Human and murine IL-7 are cross-species reactive. Recombinant Human IL-7 is a 17.4 kDa protein containing 153 amino acid residues.
- Animal-free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- 76303-576
- 76303-786
- Protein/peptide name:
- IL-7
- IL-7
- Protein synonyms:
- Lymphopoietin 1(LP-1)|Interleukin-7|pre-B cell factor
- Lymphopoietin 1(LP-1)|Interleukin-7|pre-B cell factor
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Gene ID:
- 3574
- 3574
- Sequence:
- MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH
- MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH
- Animal-free:
- Y
- Y
- Endotoxin level:
- Low
- Low
- Biological activity:
- Determined by its ability to stimulate the proliferation of mouse 2E8 cells. The expected ED50 ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of murine 2E8 cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg.
- Purity:
- 98%
- 98%
- Formulation:
- Lyophilized
- Lyophilized
- Molecular weight:
- 17.4 kDa
- 17.4 kDa
- Shipping temperature:
- Ambient
- Ambient
Spécifications
Product Family Options
Informations sur le produit
- TailleAvailabilityPrice
- Supplier Number: AF-200-07-50UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.76303-576Protein/peptide nameIL-7Protein synonymsLymphopoietin 1(LP-1)|Interleukin-7|pre-B cell factorProtein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID3574SequenceMDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEHAnimal-freeYEndotoxin levelLowBiological activityDetermined by its ability to stimulate the proliferation of mouse 2E8 cells. The expected ED50 ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg.Purity98%FormulationLyophilizedMolecular weight17.4 kDaShipping temperatureAmbient - Supplier Number: 200-07-01ML'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.76303-786Protein/peptide nameIL-7Protein synonymsLymphopoietin 1(LP-1)|Interleukin-7|pre-B cell factorProtein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID3574SequenceMDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEHAnimal-freeYEndotoxin levelLowBiological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of murine 2E8 cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg.Purity98%FormulationLyophilizedMolecular weight17.4 kDaShipping temperatureAmbient



