Human Recombinant FGF-19 (from E. coli)
À propos de cet article
The FGF family plays central roles during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-19, a member of the FGF family, is a high-affinity heparin dependent ligand for FGFR4. FGF-19 is expressed during brain development and embryogenesis. Recombinant Human FGF-19 is a 21.8 kDa protein containing 195 amino acid residues.
- Animal-free
3 Options Available Below
Product Details & Documents
The FGF family plays central roles during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-19, a member of the FGF family, is a high-affinity heparin dependent ligand for FGFR4. FGF-19 is expressed during brain development and embryogenesis. Recombinant Human FGF-19 is a 21.8 kDa protein containing 195 amino acid residues.
- Animal-free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- 76081-526
- 76081-528
- 76081-536
- Protein/peptide name:
- FGF-19
- FGF-19
- FGF-19
- Protein synonyms:
- Fibroblast Growth Factor-19|FGFJ
- Fibroblast Growth Factor-19|FGFJ
- Fibroblast Growth Factor-19|FGFJ
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 9965
- 9965
- 9965
- Sequence:
- MRPLAFSDAG PHVHYGWGDP IRLRHLYTSG PHGLSSCFLR IRADGVVDCA RGQSAHSLLE IKAVALRTVA IKGVHSVRYL CMGADGKMQG LLQYSEEDCA FEEEIRPDGY NVYRSEKHRL PVSLSSAKQR QLYKNRGFLP LSHFLPMLPM VPEEPEDLRG HLESDMFSSP LETDSMDPFG LVTGLEAVRS PSFEK
- MRPLAFSDAG PHVHYGWGDP IRLRHLYTSG PHGLSSCFLR IRADGVVDCA RGQSAHSLLE IKAVALRTVA IKGVHSVRYL CMGADGKMQG LLQYSEEDCA FEEEIRPDGY NVYRSEKHRL PVSLSSAKQR QLYKNRGFLP LSHFLPMLPM VPEEPEDLRG HLESDMFSSP LETDSMDPFG LVTGLEAVRS PSFEK
- MRPLAFSDAG PHVHYGWGDP IRLRHLYTSG PHGLSSCFLR IRADGVVDCA RGQSAHSLLE IKAVALRTVA IKGVHSVRYL CMGADGKMQG LLQYSEEDCA FEEEIRPDGY NVYRSEKHRL PVSLSSAKQR QLYKNRGFLP LSHFLPMLPM VPEEPEDLRG HLESDMFSSP LETDSMDPFG LVTGLEAVRS PSFEK
- Animal-free:
- Y
- Y
- Y
- Endotoxin level:
- Low
- Low
- Low
- Biological activity:
- Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 for this effect is 100-150 ng/ml.
- Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 for this effect is 100-150 ng/ml.
- Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 for this effect is 100-150 ng/ml.
- Purity:
- 95%
- 95%
- 95%
- Formulation:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular weight:
- 21.8 kDa
- 21.8 kDa
- 21.8 kDa
- Shipping temperature:
- Ambient
- Ambient
- Ambient
Spécifications
Product Family Options
Informations sur le produit
- TailleAvailabilityPrice
- Supplier Number: AF-100-32-5UGSpécifications:
More
Specifications
Cat. No.76081-526Protein/peptide nameFGF-19Protein synonymsFibroblast Growth Factor-19|FGFJProtein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID9965SequenceMRPLAFSDAG PHVHYGWGDP IRLRHLYTSG PHGLSSCFLR IRADGVVDCA RGQSAHSLLE IKAVALRTVA IKGVHSVRYL CMGADGKMQG LLQYSEEDCA FEEEIRPDGY NVYRSEKHRL PVSLSSAKQR QLYKNRGFLP LSHFLPMLPM VPEEPEDLRG HLESDMFSSP LETDSMDPFG LVTGLEAVRS PSFEKAnimal-freeYEndotoxin levelLowBiological activityDetermined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 for this effect is 100-150 ng/ml.Purity95%FormulationLyophilizedMolecular weight21.8 kDaShipping temperatureAmbient - Supplier Number: AF-100-32-25UGSpécifications:
More
Specifications
Cat. No.76081-528Protein/peptide nameFGF-19Protein synonymsFibroblast Growth Factor-19|FGFJProtein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID9965SequenceMRPLAFSDAG PHVHYGWGDP IRLRHLYTSG PHGLSSCFLR IRADGVVDCA RGQSAHSLLE IKAVALRTVA IKGVHSVRYL CMGADGKMQG LLQYSEEDCA FEEEIRPDGY NVYRSEKHRL PVSLSSAKQR QLYKNRGFLP LSHFLPMLPM VPEEPEDLRG HLESDMFSSP LETDSMDPFG LVTGLEAVRS PSFEKAnimal-freeYEndotoxin levelLowBiological activityDetermined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 for this effect is 100-150 ng/ml.Purity95%FormulationLyophilizedMolecular weight21.8 kDaShipping temperatureAmbient - Supplier Number: AF-100-32-1MGSpécifications:
More
Specifications
Cat. No.76081-536Protein/peptide nameFGF-19Protein synonymsFibroblast Growth Factor-19|FGFJProtein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID9965SequenceMRPLAFSDAG PHVHYGWGDP IRLRHLYTSG PHGLSSCFLR IRADGVVDCA RGQSAHSLLE IKAVALRTVA IKGVHSVRYL CMGADGKMQG LLQYSEEDCA FEEEIRPDGY NVYRSEKHRL PVSLSSAKQR QLYKNRGFLP LSHFLPMLPM VPEEPEDLRG HLESDMFSSP LETDSMDPFG LVTGLEAVRS PSFEKAnimal-freeYEndotoxin levelLowBiological activityDetermined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 for this effect is 100-150 ng/ml.Purity95%FormulationLyophilizedMolecular weight21.8 kDaShipping temperatureAmbient