Slide Holders, Jaece
Slide Holders, Jaece
Supplier:   Jaece
Slide Holders, Jaece
Supplier:  Jaece
Certificates
Slide Holders, Jaece
Slide Holders, Jaece
2 Options Available Below
Product Details & Documents
High-quality white plastic receptacle to hold slides during evaluation and cleaning.
  • Durable plastic with beveled edges
  • 39 or 78 slide capacity formats available
  • Chemical resistant
  • High-density polyethylene construction
  • Surface-griping rubber feet
These strong, lightweight slide holders are chemical resistant and provide an inexpensive way to dry slides and drain excess fluids from flat surfaces. Beveled edges provide easy access to channels and aid in slide placement or withdrawal. Inscriptions with Identi-Marker pens may be made for quick identification.
Product Family Options
Product Information
  • Désignation
    Dimensions
    No. of Places
    Availability
    Price
  • Slide Holder
    17.8×30.5 cm (7×12")
    78
    Log in for availability
    Log in for availability
    Each
    Each - $137.62
    $137.62
    Each
       $137.62
    Supplier Number: L500-D
  • Slide Holder
    10.2×30.5 cm (4×12")
    39
    Log in for availability
    Log in for availability
    Each
    Each - $106.19
    $106.19
    Each
       $106.19
    Supplier Number: L500-C