Tabletop World Relief Globe
World Relief Globe
Catalog # 470311-726
Supplier: American 3B Scientific
Fournisseur #: 1022234
Jump to:
Product Details & Documents
Tabletop globe with lighting on a metal-reinforced plastic stand with double-image map and tactile 3D relief of mountain ranges.
- Diameter: 30 cm
- Total height: 43 cm
When the globe is not lit up, it shows a physical map of the earth. When it is lit up, the current political position is shown with a contrasting delimitation of the countries and their borders.
Spécifications
- Désignation:World Relief Globe