Mini Block Heaters | Avantor
Mini Block Heaters
Mini Block Heaters
Certificates
Mini Block Heaters
Mini Block Heaters
6 Options Available Below
Product Details & Documents
Affordable mini dry-block heaters are a great option for the lab.
  • LCD Display of both temperature and time
  • Overheating protection
  • Rapid, 3-point self-calibration
  • USB Interface to save data
  • Available with or without lid/cooling
These single block mini heaters come with a block that can hold fifteen, 1.5 ml tubes, but with various additional blocks available, most common tubes can be used in these units. Three different varieties allow to choose whether or not you want to utilize a heated lid, and whether you want heating or heating and cooling. Choose the machine that best suits your needs.
Product Family Options
Blocks only
Informations sur le produit
  • Désignation
    For Tube Volume
    Availability
    Price
  • Block, fits 40 tubes
    0.2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $220.80
    $220.80
    Each
       $220.80
    Supplier Number: 18900414
  • Block, fits 24 tubes
    0.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $220.80
    $220.80
    Each
       $220.80
    Supplier Number: 18900415
  • Block, fits 15 tubes
    1.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $220.80
    $220.80
    Each
       $220.80
    Supplier Number: 18900416
  • Block, fits 15 tubes
    2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $220.80
    $220.80
    Each
       $220.80
    Supplier Number: 18900428
  • Block, fits 4 tubes
    15 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $224.55
    $224.55
    Each
       $224.55
    Supplier Number: 18900426
  • Block, fits 2 tubes
    50 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $235.50
    $235.50
    Each
       $235.50
    Supplier Number: 18900427