ContactUsLinkComponent
Adhesive Vinyl Labels 'Caution This Door Interlocked', HCL Labels
Catalog # 103727-792
Supplier:  HCL Label
undefined
Adhesive Vinyl Labels 'Caution This Door Interlocked', HCL Labels
Catalog # 103727-792
Supplier:  HCL Label
Supplier Number:  SSL0007VN00235
Specifications
  • Description:
    Preprinted adhesive labels
  • Dimensions:
    2×3"
  • Material:
    Vinyl, adhesive
  • Cat. no.:
    103727-792
  • Legend:
    Caution This Door Interlocked
Specifications
About this item
HCL's caution this door interlocked label is made of a durable adhesive vinyl.
Use this label to mark doors that are interlocked. Black and yellow coloring make this label highly visible and easy to read.
Packaging: Supplied in packs of 25.