Amino Labs Engineer-It Kit™
À propos de cet article
Full STEAM ahead with the Engineer-It Kit™; grow and engineer bacteria to create color pigments.
- Safety First! The kit uses a non-pathogenic strain of E. coli (K-12)
- No need to purchase any additional reagents
- Recommended for use with Amino Labs DNA Playground™ (available separately)
- Kit complements several Next Generation Science Standards (NGSS)
Learn genetic engineering hands-on using the Engineer-It Kit™. The end result is that you have grown and engineered bacteria to produce something for you - a colorful bio-pigment. 1-color Engineer-It Kits™ typically come with a plasmid to produce purple pigment (substitution may be made based on availability). Full Rainbow Engineer-It Kit™ comes with enough material for 10 groups and each will get their own colored plasmid.
Additional equipment required but not included: Microwave or hot-plate, and Amino Labs DNA Playground™. (In the absence of a DNA Playground™, you will also need an ice bath, water bath, thermometer, timer, and incubator).
After you’ve completed any of the Engineer-It Kits™, you can use our follow-up kits, such as the Keep-it Kit™ to save your work, or Extract-it Kit™ to break open the cells and extract your engineered creation.
Complements the Next Generation Science Standards (NGSS): MS-LS1-1, MS-LS1-5, MS-LS1-8, HS-LS1-1, HS-LS3-1, HS-LS4-4, HS-LS4-5.
Ordering information: Due to popular demand, we recommend you place your order at least a week in advance.
Delivery information: Though kits will ship at ambient temperature, some components will require refrigeration upon arrival. Refer to instructions.
4 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Full STEAM ahead with the Engineer-It Kit™; grow and engineer bacteria to create color pigments.
- Safety First! The kit uses a non-pathogenic strain of E. coli (K-12)
- No need to purchase any additional reagents
- Recommended for use with Amino Labs DNA Playground™ (available separately)
- Kit complements several Next Generation Science Standards (NGSS)
Learn genetic engineering hands-on using the Engineer-It Kit™. The end result is that you have grown and engineered bacteria to produce something for you - a colorful bio-pigment. 1-color Engineer-It Kits™ typically come with a plasmid to produce purple pigment (substitution may be made based on availability). Full Rainbow Engineer-It Kit™ comes with enough material for 10 groups and each will get their own colored plasmid.
Additional equipment required but not included: Microwave or hot-plate, and Amino Labs DNA Playground™. (In the absence of a DNA Playground™, you will also need an ice bath, water bath, thermometer, timer, and incubator).
After you’ve completed any of the Engineer-It Kits™, you can use our follow-up kits, such as the Keep-it Kit™ to save your work, or Extract-it Kit™ to break open the cells and extract your engineered creation.
Complements the Next Generation Science Standards (NGSS): MS-LS1-1, MS-LS1-5, MS-LS1-8, HS-LS1-1, HS-LS3-1, HS-LS4-4, HS-LS4-5.
Ordering information: Due to popular demand, we recommend you place your order at least a week in advance.
Delivery information: Though kits will ship at ambient temperature, some components will require refrigeration upon arrival. Refer to instructions.
Product Family Options
Informations sur le produit
- DésignationNo. of Lab GroupsAvailabilityPrice
- Supplier Number: WWE004-S_CAL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Supplier Number: WWE004-G_CAL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Supplier Number: WWE004-SR_CAL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- 1-color Engineer-It Kit™, Large Refill8Discontinued-Supplier Number: -L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Supplier Number: WWE004-F_CAL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.