IKA PETTE fix and vario, Single Channel Pipettes, IKA WORKS
Supplier: IKA Works
Certificates
About this item
An ergonomic feel combined with uncompromising precise performance: the IKA single fix and vario pipettes can be used for many applications in the modern laboratory.
- Exchangeable grips in different shapes and materials
- Tip cone and piston are coated with a high quality diamind-like carbon (DLC)
- Large display which shows set volumes in easily read characters
- User-friendly IKA software
- Simple maintenance and adjustment
Color coding simplifies the rapid selection of a suitable pipette. Resistance to impact, UV and chemicals ensures that the tip cone provides long-term reliable dosing. The PETTE fix and vario are fully autoclavable, therefore ensuring easy sterilization. This guarantees fault-free, precise and safe operation.
Frequently Bought Together