Gamma Wipe® VP Low Endotoxin Sterile Cleanroom Wipes
Supplier: Berkshire
New Product
Certificates
About this item
Gamma Wipe® VP is an ultrasonically sealed-edge cleanroom laundered wiper recommended for ISO Class 3 and above environments composed of 100% continuous filament polyester knit fabric.
- Critically low particles, fibers, ions, and extractables
- High abrasion resistance
- High absorbency
- Compatible with common cleaning and disinfecting solutions including isopropyl alcohol, phenols, bleaches, and quarternary ammonium compounds
- Lightweight, high absorbency material reduces cost in use compared to heavier weight products
Gamma Wipe® VP is composed of 100% continuous filament polyester knit fabric. Each package is gamma irradiated sterile to a 10⁻⁶ Sterility Assurance Level. This combination of properties provides Gamma Wipe® VP with the highest level of cleanliness, abrasion resistance, and chemical compatibility required for cleaning critical sterile processing environments.
Certifications: ISO class 3 - 5. Cleanroom class 1 - 100. EU Grade A/B/C/D. USP <797>.
Ordering information: Laundered and packaged in Berkshire’s ISO Class 4 cleanroom with lot number on each pack.
Frequently Bought Together