Carson PocketMicro™ Pocket Microscope | Avantor
Carson PocketMicro™ Pocket Microscope
Pocket Microscope, 20× - 60×
Catalog # 470317-028
Supplier:   CARSON OPTICAL
Fournisseur #:  MM-450
Carson PocketMicro™ Pocket Microscope
Catalog # 470317-028
Supplier:  CARSON OPTICAL
Carson PocketMicro™ Pocket Microscope
Carson PocketMicro™ Pocket Microscope
Pocket Microscope, 20× - 60×
Price
$32.85
Log in pour la disponibilité
Each
Product Details & Documents
The MM-450, PocketMicro™ is a powerful 20 to 60× power pocket microscope with an extremely lightweight and portable design. This pocket microscope features a built-in LED light that provides a bright, clear image.
It fits easily in your pocket so you can bring it with you anywhere.
Spécifications
  • Grossissement:
    20× - 60×
  • Désignation:
    Pocket Microscope