Baseline® Windmill Spirometers
Supplier: Fabrication Enterprises
Jump to:
À propos de cet article
Compact and lightweight.
- Replacement mouthpieces in plastic or paper
Compact, lightweight spirometer for screening large populations. Subject blows into disposable mouthpiece. Indicator registers forced vital lung capacity up to 7 liters.
Delivery information: Includes 50 mouthpieces.
3 Options Available Below
Product Details & Documents
Compact and lightweight.
- Replacement mouthpieces in plastic or paper
Compact, lightweight spirometer for screening large populations. Subject blows into disposable mouthpiece. Indicator registers forced vital lung capacity up to 7 liters.
Delivery information: Includes 50 mouthpieces.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Windmill SpirometerEach$359.65Case of 10$3,416.70Supplier Number: 12-1710
- Plastic MouthpiecesPack of 100$72.50Case of 1000$689.75Supplier Number: 12-1711
- Paper MouthpiecesPack of 250$81.40Case of 1000$309.25Supplier Number: 12-1714