Accessory Chuck Guard with Magnetic Enclosure, Caframo
Fournisseur: Caframo
Certificats
À propos de cet article
An accessory that adds a protective and safe guard to your spinning chuck by quickly snapping into place.
- Magnets securely snap guard into place on stirrer housing over the chuck – Easy to install and remove
- Creates a barrier around the spinning chuck – Protects users from a rotating chuck
- Chemical resistant polyurethane – Protects the chuck from splashes
Implement safety with a quick snap of this clear polyurethane chuck guard to the stirrer housing. Creates barrier around moving chuck, as well as protects chuck from chemical splashes. Easy to attach and remove.
Can be used with Caframo overhead stirrers BDC1850 (CA58960-180), BDC3030 (CA58960-165), and BDC6015 (CA58960-170).
Frequently Bought Together