Cardionics® Ausculette
Cardionics® Ausculette
Cardionics® Ausculette
Certificates
Cardionics® Ausculette
Cardionics® Ausculette
2 Options Available Below
Product Details & Documents
Great for Online Classes.
The Ausculette II is a small, battery or line-operated telemedicine auscultation system used primarily in telemedicine. It consists of 2 stethophones, a stethoscope assembly and wall-mounted power transformer.
When used with either the Tandberg or Polycom Teleconferencing System, the appropriate connecting cable is supplied.
The Ausculette can be used as both a sending and receiving auscultation system. The stethoscope is placed on the patient in the normal manner. Each listener inserts a stethophone into the rear panel of the Ausculette II and can hear simultaneously. Sounds can be amplified or filtered, if desired. A Hi-Pass filter allows attenuation of sounds from 20 Hz to 500 Hz. Heart and breath sounds can be filtered to accentuate grade I murmurs or very faint fine crackles.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Ausculette
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,142.60
    $3,142.60
    Each
       $3,142.60
    Supplier Number: 1021564
  • Stethophone
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $277.90
    $277.90
    Each
       $277.90
    Supplier Number: 1021565