Cardionics® Simulscope Bedside System
Simulscope bedside system
Catalog # 470317-918
Supplier: American 3B Scientific
Fournisseur #: 1021563
Jump to:
Product Details & Documents
Share stethoscope sounds with others in your sim center.
- Includes six heartman infrared headphones
- Excellent for bedside instruction
- Includes simulscope carrying case
- Available optional 3-Wire ECG function
The SimulScope allows clinicians and students the ability to hear live patient physiological sounds with up to 20 people simultaneously. The SimulScope is designed to provide targeted, bedside instruction for students, while minimizing intrusion on the patient. It is also beneficial in classrooms and simulation centers when auscultating simulators or manikins.
Spécifications
- Désignation:Simulscope bedside system