Cardionics® Classroom Learning Hardware | Avantor
Cardionics® Classroom Learning Hardware
Cardionics® Classroom Learning Hardware
Certificates
Cardionics® Classroom Learning Hardware
Cardionics® Classroom Learning Hardware
2 Options Available Below
Product Details & Documents
Share auscultation sounds with an entire classroom.
  • Broadcasts sounds to larger audiences
  • Excellent sound quality
  • Easy to set up and use
  • Available for large and small auditoriums
The Classroom Infrared Emitter (CIE) and Auditorium Infrared Sound System (ISS) are designed to provide simultaneous listening for groups of any size. The CIE is intended for classrooms or simulation laboratories, while the ISS is optimal for larger classrooms and auditoriums with 30-90 members. Used in conjunction with the HeartMan Infrared Headphones, audience members will be able to listen to simulated or real patient sounds alongside the presenter for clearer interaction with the sounds.
Cardionics designed the HeartMan Infrared Headphones for use with the SimulScope Auscultation System, Classroom Infrared Emitter, and Auditorium Infrared Sound System. The HeartMan Infrared Headphone works with these systems to allow simultaneous listening for clinical and educational situations.
Ordering information: The ISS And CIE Systems do not include headphones. Please see your sale rep for package options based on your needs.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Heartman Headphones
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $518.69
    $518.69
    Each
       $518.69
    Supplier Number: 1022460
  • Classroom Emitter (CIE)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,189.20
    $4,189.20
    Each
       $4,189.20
    Supplier Number: 1021982