Quick Serve Tap for Cubitainers®, Qorpak®
Catalogue # 24301-704
Fournisseur: Qorpak
Small Business Enterprise
Spécifications
- Color:White
- Description:Quick Serve Tap for Cubitainers®
- Material:HDPE
- Cat. no.:24301-704
- Cap Type:Screw thread
- Cap Size:38 mm
- Supplier No.:PLA-03193
Spécifications
À propos de cet article
This tap makes it quick and easy to pour liquids from one container to another.
- Easy-action lever tap
- Works well with Cubitainers® and vented rigid containers
Features an excellent flow rate of 72 ml/sec up to 155 ml/sec. Typical markets include beverage, liquid food, household, and chemicals.