Hula Dancer basic Orbital Shaker, IKA
À propos de cet article
The Hula Dancers are powerful orbital shakers.
- Robust housing
- High stability
- Touch function
Hula Dancer basic has a shaking diameter of 4.5 mm and allows a maximum load of 100 g. Speed is fixed at 2800 rpm.
Permissible relative humidity is 80%.
Certifications: IP protection class according to DIN EN 60529: IP 42.
2 Options Available Below
Product Details & Documents
The Hula Dancers are powerful orbital shakers.
- Robust housing
- High stability
- Touch function
Hula Dancer basic has a shaking diameter of 4.5 mm and allows a maximum load of 100 g. Speed is fixed at 2800 rpm.
Permissible relative humidity is 80%.
Certifications: IP protection class according to DIN EN 60529: IP 42.
Specifications for Product Family
Spécifications
- Catalog No:
- 76329-818
- 76329-686
- Model:
- Hula Dancer basic
- Hula Dancer digital
- Shaking motion:
- Orbital
- Orbital
- Shaking speed:
- 2800 rpm (fixed)
- 1200 - 2800 rpm (adjustable)
- Max. load:
- 0.1 kg
- 0.1 kg
- Power input/output:
- 9.5/5.0 W
- 9.5/5.0 W
- Orbit:
- 4.5 mm
- 4.5 mm
- Weight:
- 0.7 kg
- 0.7 kg
- W×D×H:
- 123×123×42 mm
- 123×123×42 mm
- Electrical:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
Spécifications
Product Family Options
Informations sur le produit
- DésignationPlage de températureAvailabilityPrice
- Supplier Number: 0025004320Spécifications:
More
Specifications
Cat. No.76329-818ModelHula Dancer basicShaking motionOrbitalShaking speed2800 rpm (fixed)Max. load0.1 kgPower input/output9.5/5.0 WOrbit4.5 mmWeight0.7 kgW×D×H123×123×42 mmElectrical100 - 240 V, 50/60 Hz - Supplier Number: 0025003634Spécifications:
More
Specifications
Cat. No.76329-686ModelHula Dancer digitalShaking motionOrbitalShaking speed1200 - 2800 rpm (adjustable)Max. load0.1 kgPower input/output9.5/5.0 WOrbit4.5 mmWeight0.7 kgW×D×H123×123×42 mmElectrical100 - 240 V, 50/60 Hz