GTS Excel Shippers, Cold Chain Technologies
Supplier: Cold Chain Technologies
Certificates
About this item
GTS Excel Shippers better ensure patient safety and regulatory compliance while costing less to procure and ship.
- Four payload sizes and three temperature ranges results in 12 total solutions
- Powered by award-winning Koolit® Advanced PCM GEL
- Error-free modular pack-out with single-step refrigerant conditioning
- Year-round performance in extreme temperatures
- Simplified ordering and inventory management
- Cost-effective for single-use shipments
- Interchangeable components across GTS Excel and Extreme product families
Recommended for use for high-value commercial and clinical trial product shipments, research and development shipments, as well as domestic expedited and ground shipments. GTS Excel Shippers are ideal for projects requiring quick off the shelf solution implementation with full qualification packages.
Frequently Bought Together