Ward's® Essentials Stereomicroscope
Stereomicroscope, Cordless, 2×, 4×, Rechargeable LED
Catalog # 470315-512
Supplier: VWR
Supplier #: SE306R-P-LED
Jump to:
Product Details & Documents
Versatile and durable, this stereomicroscope is ideal for the high school classroom.
- 20× and 40× magnifications
- Interchangeable stage plates: translucent and black/white
- Adjustable interpupillary distance
- One year limited warranty
- Rechargeable LED illumination
Features include 45° inclined viewing head, post mount that adjusts the head up or down to accommodate larger objects on stage and easy-adjust collar for tension control.
Specifications
- Désignation:Stereomicroscope, Cordless, 2×, 4×
- Illumination:Rechargeable LED