Ward's® Essentials Standard Compound Microscope
Compound Monocular Standard Microscope, Rechargeable LED
Catalog # 470315-506
Supplier: VWR
Supplier #: SFC100FLED
Jump to:
Product Details & Documents
Ideal for biological use and educational applications.
- 40×, 100× and 400× magnification
- Rechargeable LED illumination
- Inclined rotatable head
- 1 year limited warranty
This microscope model includes the most commonly specified features for standard high school instruction. The glass-coated DIN optics provide clear, crisp images and the rechargeable LED illumination offers added versatility and portability.
Specifications
- Désignation:Compound Monocular Standard Microscope
- Illumination:Rechargeable LED