Ward's® Essentials Student Compound Microscope
Compound Monocular Student Microscope, Rechargeable LED
470315-504
: VWR
: SW200
Quality performance at an economical price.
- Fully rotatable head
- 40 to 1000× with 4×, 10× and 40× obectives and 10× and 25× eyepieces
- Adjustable LED illumination
- All metal arm and base
- One year limited warranty
Designed for elementary and middle school students, this microscope boasts the same features as many advanced models, but in a 3/4 size. The rugged design with metal arm and base, carry handle and cordless capability make this microscope a practical pick for classroom and field experiments.
- Désignation:Compound Monocular Student Microscope
- Illumination:Rechargeable LED