Clear Low Density Polyethylene Pallet Top Sheet, Elkay Plastics
75848-730
- Width:1524 mm (60")
- Color:Clear
- Description:LDPE bag side gusset
- Height:190.5 mm (7.5")
- Length:1524 mm (600")
- Thickness:1.25 mm (0.00125")
- Cat. no.:75848-730
- Supplier No.:12G-303060
Top sheet covers provide added protection for shrink wrapped pallets.
- Protect items and help reduce waste
- Available in all common sizes
- Help ensure that pallet contents arrive intact
Help protect contents from water and dust as well as helping to ensure the integrity of the pallet contents.