Tuf-R® Standard Linear Low Density Gusset Bag, Elkay Plastics
Fournisseur: Elkay Plastics
Certificats
À propos de cet article
Tuf-R® Linear Low Density Polyethylene (LLDPE) bags provide extra strength and puncture resistance compared to low density bags of the same gauge.
- Tuf-R® bags can often be used in place of a heavier gauge LDPE bag to save money and reduce environmental impact
- Made from seamless tubing with no slits or side seals that can rip
- Bags are 15" wide and under come in dispenser packs of 100 for cleanliness and convenience
- Cases and inner packs are bar coded and clearly labeled for easy identification
It is often possible to use a lighter gauge Tuf-R® bag to do the job of a heavier gauge LDPE bag, which saves money and helps to reduce the environmental impact. Tuf-R® bags are made from virgin materials, so they comply with FDA requirements. Gusseted bags feature creased sides that expand to accommodate bulky items.
Certifications: Meet FDA requirements for use in food applications.
Frequently Bought Together