pH 5 to 9 Test Strips, Precision Laboratories
Catalog # 470201-152
Supplier: Precision Laboratories
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:pH Test Strips
- Cat. no.:470201-152
- pH Range:5 - 9
- Supplier No.:PH5090-1B-50
Specifications
About this item
The pH 5 - 9 test strip is a narrow range strip and is ideal for weak solutions, such as alkaline or rain water, measuring in steps of 1.0 pH units.
- Rapid test results
- Distinct color changes for easier reading and more accurate results
To use, simply dip one strip into the test solution and remove. Shake off any excess liquid. Immediately compare to the color chart for results.
Packaging: Test strips are packaged in a clear, resealable 3×4" bag.