Eppendorf® Reference® 2 Mechanical Multi-Channel Pipettes
À propos de cet article
Eppendorf® Reference® 2 multi-channel pipettes feature an innovative single-button operation, offering users precise, efficient, and ergonomic liquid handling that ensures reproducible results. Easy decontamination and a long service life will make them indispensable for high-throughput applications.
- Operated with just one button for volume selection, pipetting, and tip ejection
- 4-digit display with magnifying window for quick and secure volume setting and identification (clearly visible from every angle), incl. volume lock
- The unique smooth surface and full autoclavability guarantee efficient decontamination - making it ideal when working under sterile conditions
- Temporary adjustment allows for an easy adjustment to accurately pipette challenging liquids or in changing external conditions without the need for a full calibration
- Spring-loaded tip cones, which can be switched on/off, improve user-to-user reproducibility and ergonomics by lowering the tip attachment and ejection force
Pipetting type: air-cushion system
Operating type: mechnical
Compatible tips: epT.I.P.S.®, Universal tips
Applications include forward pipetting, reverse pipetting, removal of supernatants, sample mixing, phase extraction, and filling of plates, gels, and reaction vessels.
: All Reference® 2 pipettors are supplied with an ep.T.I.P.S. box.
5 Options Available Below
Product Details & Documents
Eppendorf® Reference® 2 multi-channel pipettes feature an innovative single-button operation, offering users precise, efficient, and ergonomic liquid handling that ensures reproducible results. Easy decontamination and a long service life will make them indispensable for high-throughput applications.
- Operated with just one button for volume selection, pipetting, and tip ejection
- 4-digit display with magnifying window for quick and secure volume setting and identification (clearly visible from every angle), incl. volume lock
- The unique smooth surface and full autoclavability guarantee efficient decontamination - making it ideal when working under sterile conditions
- Temporary adjustment allows for an easy adjustment to accurately pipette challenging liquids or in changing external conditions without the need for a full calibration
- Spring-loaded tip cones, which can be switched on/off, improve user-to-user reproducibility and ergonomics by lowering the tip attachment and ejection force
Pipetting type: air-cushion system
Operating type: mechnical
Compatible tips: epT.I.P.S.®, Universal tips
Applications include forward pipetting, reverse pipetting, removal of supernatants, sample mixing, phase extraction, and filling of plates, gels, and reaction vessels.
: All Reference® 2 pipettors are supplied with an ep.T.I.P.S. box.
Specifications for Product Family
Spécifications
- Catalog No:
- 10032-658
- CA10032-724
- CA10032-520
- CA10032-660
- CA10032-658
- CA10032-662
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
Spécifications
Product Family Options
Informations sur le produit
- DésignationVolumeIncrementsColor CodeNo. of ChannelsAvailabilityPrice
- Reference® 2, 12-channel, variable volume, spring-loaded tip0.5 - 10 µl0.01 µlLight orange12Discontinued-Supplier Number: 4926000026Spécifications:
Low Manufacturing Impact More
Specifications
Cat. No.10032-658Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Reference® 2, 8-channel, variable volume, spring-loaded tip0.5 - 10 µl0.01 µlLight orange8Log in pour la disponibilitéEach$2,260.00Supplier Number: 4926000018Spécifications:
Low Manufacturing Impact More
Specifications
Cat. No.CA10032-724Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Reference® 2, 8-channel, variable volume, spring-loaded tip10 - 100 µl0.1 µlYellow8Log in pour la disponibilitéEach$2,260.00Supplier Number: 4926000034Spécifications:
Low Manufacturing Impact More
Specifications
Cat. No.CA10032-520Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Reference® 2, 12-channel, variable volume, spring-loaded tip10 - 100 µl0.1 µlYellow12Log in pour la disponibilitéEach$2,670.00Supplier Number: 4926000042Spécifications:
Low Manufacturing Impact More
Specifications
Cat. No.CA10032-660Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Reference® 2, 12-channel, variable volume, spring-loaded tip0.5 - 10 µl0.01 µlMedium gray12Log in pour la disponibilitéEach$2,260.00Supplier Number: 4926000050Spécifications:
More
Specifications
Cat. No.CA10032-658Environmentally PreferableEnvironmental certification - Reference® 2, 12-channel, variable volume, spring-loaded tip30 - 300 µl0.2 µlOrange12Log in pour la disponibilitéEach$2,670.00Supplier Number: 4926000069Spécifications:
Low Manufacturing Impact More
Specifications
Cat. No.CA10032-662Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact
Accessories
Informations sur le produit
- Catalog #CapacityAvailabilityPrice
- Reagent reservoir tip-tub
Informations sur le produit
- Catalog #AvailabilityPrice
- Pipette carousel 2