Cryogenic Vials | Avantor
Cryogenic Vials
Supplier:   GSC International
Cryogenic Vials
Supplier:  GSC International
Certificates
Cryogenic Vials
2 Options Available Below
Product Details & Documents
Cryogenic vials are used for storing biological samples at extreme temperatures.
  • Leak-proof
  • Printed graduations
  • White area for labeling
Our vials can withstand temperatures as low as –196 °C (approximately –321 °F) and have leak-proof silicone rubber lined caps, printed graduations, and a white area for labeling their contents.
1.8 ml vials measure 45.7 mm (1.8") tall and 13 mm (0.51") in diameter, and our 5 ml vials measure 56.9 mm (2.24") tall and 20 mm (0.79") in diameter.
Product Family Options
Informations sur le produit
  • Capacité
    Diameter ×H
    Avec bouchon
    Availability
    Price
  • 1.80 ml
    13×45.7 mm
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 500
    Pack of 500 - $185.35
    $185.35
    Log in pour la disponibilité
    Case of 2000
    Case of 2000 - $740.85
    $740.85
    Pack of 500
       $185.35
    Case of 2000
       $740.85
    Supplier Number: CRYTB-1.8
  • 5.00 ml
    20×56.9 mm
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 200
    Pack of 200 - $119.75
    $119.75
    Pack of 200
       $119.75
    Supplier Number: CRYTB-5