TLC Plates, Cellulose MN 300, Macherey-Nagel
Fournisseur: Macherey-Nagel
Certificats
À propos de cet article
Fibrous cellulose layers are available as POLYGRAM® polyester sheets.
- Alternative selectivity to silica 60
- Fiber length (95%) 2 to 20 μm
- Average degree of polymerization 400 to 500
- Specific surface according to Blaine 15000 cm2/g
- ≤20 ppm Fe, 6 ppm Cu, 7 ppm P
- Dichloromethane extract ≤0.25%
- Residue on ignition at 850 °C ≤1500 ppm
They are used in partition chromatography of polar substances such as for the separations of amino acids, carboxylic acids or carbohydrates.
Frequently Bought Together