Bacterial Spreader
Bacterial Spreader, Stainless steel
Catalog # 470315-230
Supplier: DR INSTRUMENTS INC. SE
Supplier #: BTS9
Jump to:
Product Details & Documents
For fast and easy plating.
- Rounded to minimize damage to the plate
For simple and fast spreading of suspensions onto agar plates.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Désignation:Bacterial Spreader
- Matière:Stainless steel