ContactUsLinkComponent
Featherweight Forceps Set
Catalogue # 470315-238
Fournisseur:  DR INSTRUMENTS INC. SE
undefined
Featherweight Forceps Set
Catalogue # 470315-238
Fournisseur:  DR INSTRUMENTS INC. SE
Supplier Number:  DRENTF-II
Spécifications
  • Description:
    Featherweight Forceps Set
  • Cat. no.:
    470315-238
Spécifications
À propos de cet article
Use a soft touch for delicate specimens.
  • Light closure for delicate specimens
Featherweight entomology forceps set with one narrow tip and one wide tip forceps. Made out of flexible stainless steel (Spring Stainless Steel), these forceps can be bent virtually in any shape and form. These forceps are ideal for handling larvae or eggs without crushing them and specimens stored in vials.
Delivery: Set includes broad and fine points and plastic storage case.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.