ADC® Prosphyg 768 Pocket Aneroid Sphygmomanometer
Catalog # 470315-708
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Color:Black
- Description:Prosphyg 768
- Size:Adult (22 - 32 cm)
- Cat. no.:470315-708
Specifications
About this item
Affordable and professional.
- Zippered color matched case for storage
- Not made with natural rubber latex
ADC®'s prosphyg 768 series is our most popular sphygmomanometer. From the contemporary black enamel manometer to the meticulously designed adcuff inflation system, the 768 is the model of choice for professionals looking for high quality at an affordable price.
Delivery: Includes 300 MMHG manometer, extra-large, color-matched, dual-pocket nylon carry case.