ADC® Calibration Test Kit
Calibration Test Kit
Catalog # 470315-738
Supplier: AMERICAN DIAGNOSTIC CORP CP
Fournisseur #: 991
Jump to:
Product Details & Documents
Make sure your gauges are calibrated.
- Not made of natural rubber latex
Do you think you have a manometer that needs calibration? Easily check to see if two devices agree using this simple calibration test kit. Simply connect a known good device, and a device in question, to the Y tubing, and apply pressure using the bulb. Both devices should give the same reading. If they don't you definitely need recalibration.
Delivery information: Includes connectors to permit testing of aneroid and mercury sphygs.
Specifications
- Désignation:Calibration Test Kit