ADC® Multikuf 3 Cuff Sphygmomanometer | Avantor
ADC® Multikuf 3 Cuff Sphygmomanometer
Portable Multicuff Kit, Black
Catalog # 470315-702
Fournisseur #:  731-BK
ADC® Multikuf 3 Cuff Sphygmomanometer
Catalog # 470315-702
ADC® Multikuf 3 Cuff Sphygmomanometer
ADC® Multikuf 3 Cuff Sphygmomanometer
Portable Multicuff Kit, Black
Price
$346.40
Log in pour la disponibilité
Each Case of 5
La quantité minimale de commande est de 1
Product Details & Documents
Multiple cuff access in a compact case.
  • Durable palm aneroid
  • Compact storage case
  • Not made with natural rubber latex
If you want easy flexibility in a multi-cuff set, this is a great option. The ADC® 804 palm aneroid is a precision crafted gauge in a rugged ABS housing. To further protect the instrument, it is fully contained in a zinc safety cage. The release valve is an ambidextrous trigger, so its easy to use from either hand, even while wearing gloves. Each cuff is stored in an individually labeled pocket for quick access. The folding pockets, and palm aneroid, are easily stored in a zippered case ( 8×6×3.5") for instant accessibility.
Delivery information: The set includes three sizes of adult sized, single tube, cuffs (small adult, adult and large adult) which quickly connect to the aneroid with a bayonet connection.
Spécifications
  • Color:
    Black
  • Désignation:
    Portable Multicuff Kit