ADC® Adcuff Replacement Cuff With Bladders
ADC® Adcuff Replacement Cuff With Bladders
Certificates
ADC® Adcuff Replacement Cuff With Bladders
ADC® Adcuff Replacement Cuff With Bladders
12 Options Available Below
Product Details & Documents
For use with most pocket and clock aneroids.
  • Available for single or double tube systems
  • Long 55 cm tubing
  • Three year warranty
  • Not made with natural rubber latex
Used in virtually all ADC® prosphyg and diagnostix brand instruments, the newly redesigned adcuff is the most feature rich bladdered cuff on the market. Manufactured to our exacting specifications, ADC® starts with the finest 210 denier nylon. Sewn with only nylon thread to resist puckering during laundering. The custom manufactured hook and loop adhesive is specially matched to provide a minimum of 30,000 open and close cycles. Many lesser cuffs won’t even make it to the industry mandated 10,000. ADC® randomly tests it on specialized test equipment in our facility to ensure long wear.
It feature an improved size guide marking system to reduce the risk of mis-cuffing. The new size icon system simplifies cuff selection. All ADC® cuffs now feature our bladder flap that secures the bladderand prevents “bulging”. All ADC® sphygs are now completely latex free. ADC still uses a traditional bladder. Two years has been spent testing and evaluating materials ultimately selecting a specially formulated PVC that has tensile properties similar to latex with superior environmental stability.
ADC® has tested these bladders successfully to a minimum of 50,000 cycles - 5× more than required under the ANSI SP 10 standard and 60% more than our previous latex and neoprene bladders. Each and every bladder is leak tested at the factory and we random leak test bladders a second time in our facility.
Product Family Options
Single Tubes
Informations sur le produit
  • Taille
    Color
    Availability
    Price
  • Infant (9 - 14 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $29.25
    $29.25
    Log in pour la disponibilité
    Case of 15
    Case of 15 - $416.80
    $416.80
    Each
       $29.25
    Case of 15
       $416.80
    Supplier Number: 845-7IBK-1
  • Child (13 - 19.5 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $36.35
    $36.35
    Log in pour la disponibilité
    Case of 15
    Case of 15 - $517.65
    $517.65
    Each
       $36.35
    Case of 15
       $517.65
    Supplier Number: 845-9CBK-1
  • Small Adult (19 - 27 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.75
    $39.75
    Log in pour la disponibilité
    Case of 15
    Case of 15 - $566.25
    $566.25
    Each
       $39.75
    Case of 15
       $566.25
    Supplier Number: 845-10SABK-1
  • Adult (22 - 32 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $50.20
    $50.20
    Log in pour la disponibilité
    Case of 15
    Case of 15 - $753.00
    $753.00
    Each
       $50.20
    Case of 15
       $753.00
    Supplier Number: 845-11ABK-1
  • Large Adult (34 - 50 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $65.90
    $65.90
    Log in pour la disponibilité
    Case of 15
    Case of 15 - $988.50
    $988.50
    Each
       $65.90
    Case of 15
       $988.50
    Supplier Number: 845-12XBK-1
  • Thigh (40 - 66 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $70.45
    $70.45
    Log in pour la disponibilité
    Case of 8
    Case of 8 - $535.40
    $535.40
    Each
       $70.45
    Case of 8
       $535.40
    Supplier Number: 845-13TBK-1
Two Tubes
Informations sur le produit
  • Taille
    Color
    Availability
    Price
  • Infant (9 - 14 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $29.25
    $29.25
    Log in pour la disponibilité
    Case of 15
    Case of 15 - $416.80
    $416.80
    Each
       $29.25
    Case of 15
       $416.80
    Supplier Number: 845-7IBK-2
  • Child (13 - 19.5 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.90
    $31.90
    Log in pour la disponibilité
    Case of 15
    Case of 15 - $454.60
    $454.60
    Each
       $31.90
    Case of 15
       $454.60
    Supplier Number: 845-9CBK-2
  • Small Adult (19 - 27 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.75
    $39.75
    Log in pour la disponibilité
    Case of 15
    Case of 15 - $566.25
    $566.25
    Each
       $39.75
    Case of 15
       $566.25
    Supplier Number: 845-10SABK-2
  • Adult (22 - 32 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $50.40
    $50.40
    Log in pour la disponibilité
    Case of 15
    Case of 15 - $756.00
    $756.00
    Each
       $50.40
    Case of 15
       $756.00
    Supplier Number: 845-11ABK-2
  • Large Adult (34 - 50 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $71.40
    $71.40
    Log in pour la disponibilité
    Case of 15
    Case of 15 - $1,071.00
    $1,071.00
    Each
       $71.40
    Case of 15
       $1,071.00
    Supplier Number: 845-12XBK-2
  • Thigh (40 - 66 cm)
    Black
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $80.00
    $80.00
    Log in pour la disponibilité
    Case of 8
    Case of 8 - $640.00
    $640.00
    Each
       $80.00
    Case of 8
       $640.00
    Supplier Number: 845-13TBK-2