ADC® Adscope® 641 Sprague Stethoscopes
Adscope 641, Black
Catalog # 470315-686
Supplier: AMERICAN DIAGNOSTIC CORP CP
Fournisseur #: 641BK
Jump to:
Product Details & Documents
Why own just a sprague stethoscope when you can own an adscope sprague?
- Proprietary valve mechanism prevents acoustic leakage
- Not made with natural rubber latex
The legendary design features of the Sprague have made it one of the most popular stethoscopes in history. ADC®'s Adscope Sprague 641 offers a double-tube design for improved stereo separation, with a 22" tubing length (and a 30" length overall). The threaded chest piece drum has five interchangeable fittings for complete diagnostic capability on most patients.
Delivery information: Includes accessory pouch for spare chest pieces and bells.
Spécifications
- Color:Black
- Désignation:Adscope 641